Anti-ISG15 Rabbit Monoclonal Antibody [clone: 1H12]

Supplier: Rockland Immunochemicals
200-401-438 200-401-438
RL200-401-438EA 569.75 USD
CA200-401-438 RL200-401-438
Anti-ISG15 Rabbit Monoclonal Antibody [clone: 1H12]
Antibodies
This purified polyclonal antibody reacts with human ISG15 by western blot and ELISA. Although not tested, this antibody is likely functional in immunochemistry and immunoprecipitation.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:200 - 1:1000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: ISG15
Clonality: Monoclonal
Clone: 1H12
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR