Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 14H5]

Supplier: Rockland Immunochemical
109-401-304
This antiserum against anti-Human IL-1 Receptor Type I (IL-1RI) has been tested for use in ELISA,  radioimmunoassays and immunoprecipitation.  Reactivity in other immunoassays is unknown.

Recommended Dilutions:

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: 14H5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primates




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
IL1R1, Antiserum 1 mL 109-401-304 RL109-401-304 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol IL1R1
Antigen Name Interleukin 1 Receptor
Reactivity Primates, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Clone
Host Rabbit
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes
Neutralization Yes
Antigen Synonyms IL1RA, IL-1R-alpha, IL1R, D2S1473, P80, CD121A
Gene ID 3554
Concentration 84 mg/ml by Refractometry
Format Antiserum
Cross Adsorption No
Immunogen IL-1 Receptor Type I Antibody was prepared by repeated immunizations with recombinant human IL-1R type I.
Form Liquid (sterile filtered)
Storage Buffer 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Dry Ice
Tested Applications Radio Immunoassay
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty