Anti-IgG Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-1104 109-1104
RL109-1104EA 174.82 USD
RL109-1104 CA109-1104
Anti-IgG Goat Polyclonal Antibody
Antibodies
suitable for classical and molecular immunological methods.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR