Anti-ERBB3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-205 200-401-205
RL200-401-205EA 487.39 USD
CA200-401-205 RL200-401-205
Anti-ERBB3 Rabbit Polyclonal Antibody
Antibodies
Anti-HER3 antibody is specifically designed for immunoprecipitation and can be used for immunoblotting in conjunction with a detection antibody like anti-phosphotyrosine.  Reactivity in other assays is likely, but has not been determined.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunoprecipitation: Use 5 µl antibody.;

Type: Primary
Antigen: ERBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR