Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CSF2 Rabbit Monoclonal Antibody [clone: 7A11]

Supplier: Rockland Immunochemical
109-401-314
This antiserum against Human GM-CSF has been tested for use in neutralizations, ELISA and immunoblotting. Reactivity is also expected in neutralizations and radioimmunoassay.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 7A11
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity:




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mL 109-401-314 RL109-401-314 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CSF2
Antigen Name Colony Stimulating Factor 2
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 7A11
Host Rabbit
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Neutralization Yes
Antigen Synonyms MGI-IGM, TUBB, GMCSF, Gm-CSf, Csfgm, CDCBM5, TUBB2
Gene ID 1437
Epitope Full length
Concentration 85 mg/ml by Refractometry
Format Antiserum
Cross Adsorption No
Immunogen This whole rabbit serum was prepared by repeated immunizations with full length recombinant human GM-CSF.
Form Liquid (sterile filtered)
Storage Temperature -20°C
Shipping Temperature Dry Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty