Anti-CUL4A Rabbit Monoclonal Antibody [clone: 8A3]

Supplier: Rockland Immunochemicals
100-401-A04 100-401-A04
RL100-401-A04EA 468.04 USD
RL100-401-A04 CA100-401-A04
Anti-CUL4A Rabbit Monoclonal Antibody [clone: 8A3]
Antibodies
This antibody reacts with human Cul4A by western blot and immunoprecipitation.  The antibody immunoprecipitates Protein from cell lysates (using HeLa, U2OS and others).  This antibody also co-immunoprecipitates at least 8 subunits of COP9, DDB1 and CAND1.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CUL4A
Clonality: Monoclonal
Clone: 8A3
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR