Anti-ANAPC2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-A16 100-401-A16
RL100-401-A16EA 518.16 USD
RL100-401-A16 CA100-401-A16
Anti-ANAPC2 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human APC2 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from overexpressing cell lysates (using HeLa and NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: ANAPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR