Anti-H5N1 Mouse Polyclonal Antibody [clone: 3G2]

Supplier: Rockland Immunochemicals
200-301-976 200-301-976
RL200-301-976EA 528.35 USD
RL200-301-976 CA200-301-976
Anti-H5N1 Mouse Polyclonal Antibody [clone: 3G2]
Antibodies
Hemagglutinin of A/Vietnam/1203/04 Influenza Virus (VN04-8) monoclonal antibody can be used for hemagglutination inhibition (HI) assays to provide antigenic characterization of the influenza A viruses of the H5 HA subtype.

Recommended Dilutions: ELISA: 1:5000;

Type: Primary
Antigen: H5N1 (VN04-8)
Clonality: Polyclonal
Clone: 3G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: H5N1 virus, Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR