Anti-H5N1 Mouse Polyclonal Antibody [clone: 14C5]

Supplier: Rockland Immunochemicals
200-301-979 200-301-979
RL200-301-979EA 528.35 USD
RL200-301-979 CA200-301-979
Anti-H5N1 Mouse Polyclonal Antibody [clone: 14C5]
Antibodies
Hemagglutinin of A/Vietnam/1203/04 Influenza Virus (VN04-13) monoclonal antibody can be used for hemagglutination inhibition (HI) assays to provide antigenic characterization of the influenza A viruses of the H5 HA subtype.

Recommended Dilutions: ELISA: 1:5000;

Type: Primary
Antigen: H5N1 (VN04-13)
Clonality: Polyclonal
Clone: 14C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: H5N1 virus, Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR