Anti-GST Mouse Polyclonal Antibody [clone: 3D4]

Supplier: Rockland Immunochemicals
200-301-200 200-301-200
RL200-301-200EA 580.43 USD
RL200-301-200 CA200-301-200
Anti-GST Mouse Polyclonal Antibody [clone: 3D4]
Antibodies
Primary Mouse Anti-Glutathione-S-Transferase (GST) Reacts with Pan

Type: Tag
Antigen: GST
Clonality: Monoclonal
Clone: 3D4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR