Anti-IgG Mouse Polyclonal Antibody

Supplier: Rockland Immunochemicals
105-3102 105-3102
RL105-3102EA 154.28 USD
CA105-3102 RL105-3102
Anti-IgG Mouse Polyclonal Antibody
Antibodies
Secondary Mouse Anti-IgG (H&L) Reacts with Goat

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Mouse
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR