Anti-GST Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-101-200 100-101-200
RL100-101-200EA 487.47 USD
CA100-101-200 RL100-101-200
Anti-GST Goat Polyclonal Antibody
Antibodies
Primary Goat Anti-GLUTATHIONE-S-TRANSFERASE (Schistosoma japonicum) Reacts with Human

Type: Primary
Antigen: Glutathion-S-Transferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR