Anti-GLUD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-4158 100-4158
RL100-4158EA 1289.92 USD
RL100-4158 CA100-4158
Anti-GLUD1 Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Bovine glutamate dehydrogenase exists as a homohexamer located within the mitochondrial matrix.

Recommended Dilutions: ELISA: 1:4000 - 1:16000; Western Blot: 1:1000 - 1:3000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: GLUD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR