Anti-CCNT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-158 100-401-158
RL100-401-158EA 529.5 USD
RL100-401-158 CA100-401-158
Anti-CCNT1 Rabbit Polyclonal Antibody
Antibodies
Suitable for ELISA, immunoprecipitation, immunochemistry, immunoblotting and other immunological methods requiring high titer and specificity.  Anti-Cyclin T1 is suitable for the detection by immunoblot of human, Rat and mouse Cyclin T1.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CCNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR