Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCNL1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-881
This Protein A purified antibody has been tested for use in ELISA, immunochemistry western blot, immunoprecipitation and immunofluores-cence.   Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 59 kDa in size.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 10 mg/ml - 20 µg/ml; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: CCNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Cyclin L1[alpha] (314–369), Polyclonal 500 µg 200-401-881 RL200-401-881 Each (500µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CCNL1
Antigen Name Cyclin L1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Antigen Synonyms ANIA6A, Cyclin-L1, CCNL1, UNQ530/PRO1073, BM-001, Cyclin-L, ania-6a, PRO1073
Amino Acid Number 314 to 369
Gene ID 57018
Concentration 5.0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross Adsorption No
Immunogen This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic polypeptide corresponding to amino acids 314-369 of Human Cyclin L1a Protein.
Purification Protein A purified
Form Lyophilized
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Ambient
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty