Anti-CCNB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-152 100-401-152
RL100-401-152EA 518.16 USD
CA100-401-152 RL100-401-152
Anti-CCNB1 Rabbit Polyclonal Antibody
Antibodies
This antibody is suitable for use in ELISA, immunoblotting, immunoprecipitation and other immuno-logical methods requiring high titer and specificity.   Specific conditions for reactivity and detection of Cyclin B1 should be optimized by the researcher.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CCNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR