Anti-CCNA2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-151 100-401-151
RL100-401-151EA 518.16 USD
RL100-401-151 CA100-401-151
Anti-CCNA2 Rabbit Polyclonal Antibody
Antibodies
Anti-Cyclin-A antibody is suitable for the detection by immunoblot, ELISA and IP of human, Rat and mouse Cyclin A.  Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CCNA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR