Anti-Choline oxidase Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-101-094 100-101-094
RL100-101-094EA 1289.92 USD
RL100-101-094 CA100-101-094
Anti-Choline oxidase Goat Polyclonal Antibody
Antibodies
This product has been assayed against 1.0 µg of Choline Oxidase (Alcaligenes species) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Choline oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR