Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
103-4104 103-4104
RL103-4104EA 176.89 USD
RL103-4104 CA103-4104
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Secondary Rabbit Anti-IgY F(ab')2 Reacts with Chicken

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR