Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDK9 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
This antibody has been tested for use in ELISA, immunoprecipitation, immunochemistry and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 43 kDa corresponding to CDK9 (PITALRE).

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK9
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Host: Rabbit

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 100-401-167 RL100-401-167 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol CDK9
Antigen Name Cyclin-dependent Kinase 9
Reactivity Mouse, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Western Blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Antigen Synonyms serine/threonine-protein kinase PITALRE, CTK1, RP11-228B15.5, TAK, serine/threonine protein kinase PITALRE, C-2k, tat-associated kinase complex catalytic subunit, CDC2L4, PITALRE, Cell Division Cycle 2-like protein kinase 4, CDC2-related kinase, cell division protein kinase 9
Gene ID 1025
Format Antiserum
Cross Adsorption No
Immunogen Multiple synthetic peptides corresponding to C-terminal and N-terminal domains of the Protein coded by the human gene cdk9 (PITALRE).
Purification delipidation and defibrination
Form Liquid (sterile filtered)
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Dry Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty