Anti-CDK5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-163 100-401-163
RL100-401-163EA 518.16 USD
CA100-401-163 RL100-401-163
Anti-CDK5 Rabbit Polyclonal Antibody
Antibodies
This product was assayed by immunoblot and found to be reactive against cdk5 (p31) at a dilution of 1:500 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK5
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR