Anti-CDK1 Rabbit Polyclonal Antibody [clone: 10H11.E12]

Supplier: Rockland Immunochemicals
100-401-160 100-401-160
RL100-401-160EA 518.16 USD
RL100-401-160 CA100-401-160
Anti-CDK1 Rabbit Polyclonal Antibody [clone: 10H11.E12]
Antibodies
This antibody is suitable for immunoblotting, immunochemistry immuno-precipitation (as active kinase), and immunoblotting.  HeLa cell lysate or human colon carcinoma is suggested as a positive control for immunoblotting.

Recommended Dilutions: ELISA: 1:60,000; Western Blot: 1:250 - 1:5000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR