Anti-CAT Rabbit Monoclonal Antibody [clone: 7C10D8]

Supplier: Rockland Immunochemicals
100-4151 100-4151
RL100-4151EA 1289.92 USD
RL100-4151 CA100-4151
Anti-CAT Rabbit Monoclonal Antibody [clone: 7C10D8]
Antibodies
Anti-Catalase Antibody is suitable for western blotting, Immunochemistryand for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:100000 - 1:1000,000; Western Blot: 1:5000 - 1:20000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: CAT
Clonality: Monoclonal
Clone: 7C10D8
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR