Anti-ALPI Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-142 100-401-142
RL100-401-142EA 1289.92 USD
CA100-401-142 RL100-401-142
Anti-ALPI Rabbit Polyclonal Antibody
Antibodies
This product has been assayed against 1.0 µg of Alkaline Phosphatase (Human Intestine) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR