Anti-ALPI Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-1135 100-1135
RL100-1135EA 1289.92 USD
CA100-1135 RL100-1135
Anti-ALPI Goat Polyclonal Antibody
Antibodies
Primary Goat Anti-ALKALINE PHOSPHATASE (Calf Intestine) Reacts with Pan

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR