Anti-AGAP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-946 200-401-946
RL200-401-946EA 518.16 USD
RL200-401-946 CARL200401-946
Anti-AGAP2 Rabbit Polyclonal Antibody
Antibodies
This Protein A purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 90 kDa in size corresponding to AGAP2 Protein by western blotting.

  • Recommended Dilutions: ELISA: 1:40000 - 1:200,000; Western Blot: 1:5000 - 1:20000;
  • Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: AGAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR