Anti-AMFR Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2162B
80088-444EA 751.46 USD
80088-444
Anti-AMFR Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-AMFR (C-term) Reacts with Human

Type: Primary
Antigen: AMFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (200–300 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR