Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Human Igκ Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemical
709-1510
This product has been assayed against 1.0 µg of Human IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
500 µg 709-1510 RL709-1510 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol Igκ
Antigen Name Immunoglobulin Kappa
Reactivity Human
Conjugation AP (Alkaline Phosphatase)
Clonality Polyclonal
Antibody Type Secondary
Host Goat
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope Light Chain
Concentration 1.0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross Adsorption No
Immunogen Human kappa light chain
Purification purified by immunoaffinity chromatography
Form Liquid (sterile filtered)
Antibody Fragment F(ab')2
Storage Buffer 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
Storage Temperature 4°C
Shipping Temperature Wet Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty