Anti-IgG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemicals
602-4503 602-4503
RL602-4503EA 271.52 USD
RL602-4503 CA602-4503
Anti-IgG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Secondary Rabbit Anti-IgG F(c) Reacts with Cat (Feline)

Recommended Dilutions: ELISA: 1:4000 - 1:16000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR