|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | IgG |
Antigen Name | Immunoglobulin G |
Reactivity | Cat |
Conjugation | AP (Alkaline Phosphatase) |
Clonality | Polyclonal |
Antibody Type | Secondary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
Epitope | Heavy and Light chain |
Concentration | 1.0 mg/ml by UV absorbance at 280 nm |
Format | Antibody fraction |
Cross Adsorption | No |
Immunogen | Cat IgG whole molecule |
Purification | purified by immunoaffinity chromatography |
Form | Liquid (sterile filtered) |
Storage Buffer | 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free. |
Storage Temperature | 4°C |
Shipping Temperature | Wet Ice |