Anti-NCOA3 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP1082A
60941-204EA 751.67 USD
60941-204
Anti-NCOA3 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-AIB1 (RAC3/NCOA3) (Center) Reacts with Human

Type: Primary
Antigen: NCOA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR