Anti-IgG Goat Polyclonal Antibody (Agarose)

Supplier: Rockland Immunochemicals
207-1150 207-1150
RL207-1150EA 378.42 USD
RL207-1150 CA207-1150
Anti-IgG Goat Polyclonal Antibody (Agarose)
Antibodies
Secondary Goat Anti-IgG (H&L) Reacts with Hamster

Recommended Dilutions:
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Golden Syrian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR