Anti-IgG Goat Polyclonal Antibody (Agarose)

Supplier: Rockland Immunochemicals
203-1150 203-1150
RL203-1150EA 329.19 USD
CA203-1150 RL203-1150
Anti-IgG Goat Polyclonal Antibody (Agarose)
Antibodies
Secondary Goat Anti-IgY (H&L) Reacts with Chicken

Recommended Dilutions:
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR