Anti-RAD9 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-468 600-401-468
RL600-401-468EA 518.16 USD
RL600-401-468 CA600-401-468
Anti-RAD9 Rabbit Polyclonal Antibody
Antibodies
This phospho specific polyclonal antibody was tested by immunoblotting and ELISA.  Data from both immunoblotting and ELISA indicate the antibody is reactive with the phosphorylated form of the immunizing peptide and minimally reactive with the non-phospho

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:500- 1:2000;

Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR