Anti-V5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-378 600-401-378
RL600-401-378EA 336.2 USD
CA600-401-378 RL600-401-378
Anti-V5 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-V5 EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: V5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR