Anti-TLK1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-469 600-401-469
RL600-401-469EA 518.16 USD
RL600-401-469 CA600-401-469
Anti-TLK1 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 86 kDa, 89 kDa or 64 kDa in size corresponding to isoforms of TLk-1.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: TLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Dog, Human, orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR