Anti-MER2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-924 600-401-924
RL600-401-924EA 518.16 USD
RL600-401-924 CA600-401-924
Anti-MER2 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 50 kDa in size.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:1000 - 1:10000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: MER2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S.cerevisiae, Saccharomyces cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR