Anti-ROBO1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-692 600-401-692
RL600-401-692EA 522.84 USD
RL600-401-692 CA600-401-692
Anti-ROBO1 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA , IP, immunochemistry, immunofluorescence, and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band at ~181 kDa in size.

Recommended Dilutions: ELISA: 1:30,000 - 1:160,000; Western Blot: 1:500 - 1:3000; Immunochemistry: 2 µg/ml to 10 µg/ml

Type: Primary
Antigen: ROBO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR