Anti-RFP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-379 600-401-379
RL600-401-379EA 437 USD
RL600-401-379 CA600-401-379
Anti-RFP Rabbit Polyclonal Antibody
Antibodies
Polyclonal anti-RFP is designed to detect RFP and its variants. This antibody can be used to detect RFP by ELISA (sandwich or capture) for the direct binding of antigen.

Recommended Dilutions: ELISA: 1:50000 - 1:100000; Immunofluorescence: 1:200-1:2000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:200 - 1:2000;

Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR