Anti-IgM µ Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-1107 612-1107
RL612-1107EA 189.22 USD
CA612-1107 RL612-1107
Anti-IgM µ Goat Polyclonal Antibody
Antibodies
Anti-Rat IgM (mu chain) (GOAT) Antibody has been assayed against Rat IgM in a standard capture ELISA using Peroxidase Conjugated Streptavidin. A working dilution of 1:15,000 to 1:70,000 of the concentration is suggested for this product.

Recommended Dilutions: ELISA: 1:80,000; Western Blot: 1:1000 to 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR