Anti-IgG Donkey Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-701-120 612-701-120
RL612-701-120EA 183.1 USD
RL612-701-120 CA612-701-120
Anti-IgG Donkey Polyclonal Antibody
Antibodies
Secondary Donkey Anti-IgG (H&L) Reacts with Rat

Recommended Dilutions: ELISA: 1:60,000 - 1:300,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR