Anti-IgG Donkey Polyclonal Antibody

Supplier: Rockland Immunochemicals
611-701-127 611-701-127
RL611-701-127EA 183.1 USD
RL611-701-127 CA611-701-127
Anti-IgG Donkey Polyclonal Antibody
Antibodies
Anti-Rabbit IgG (H&l) is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR