Anti-PPARD Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-420 600-401-420
RL600-401-420EA 518.16 USD
CA600-401-420 RL600-401-420
Anti-PPARD Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a single band approximately 43 kDa in size corresponding to PPAR delta by western blot.

Recommended Dilutions: ELISA: 1:600,000; Western Blot: 1:500 - 1:5000;

Type: Primary
Antigen: PPARD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR