Anti-TRF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-255 600-401-255
RL600-401-255EA 1289.92 USD
CA600-401-255 RL600-401-255
Anti-TRF Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-MOUSE TRANSFERRIN Reacts with Mouse

Recommended Dilutions:

Type: Primary
Antigen: TRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR