Anti-MDM2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-423 600-401-423
RL600-401-423EA 518.16 USD
RL600-401-423 CA600-401-423
Anti-MDM2 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user.  Expect bands approximately 102 kDa in size corresponding to phosphorylated MDM2 Protein by western blotting.

Recommended Dilutions: ELISA: 1:3000 - 1:12000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: MDM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR