Anti-IgG1 gamma Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
610-4140 610-4140
RL610-4140EA 209.83 USD
RL610-4140 CA610-4140
Anti-IgG1 gamma Rabbit Polyclonal Antibody
Antibodies
Secondary Rabbit Anti- IgG1 Reacts with Mouse

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG1 gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR