Anti-IgG Donkey Polyclonal Antibody

Supplier: Rockland Immunochemicals
610-701-124 610-701-124
RL610-701-124EA 184.76 USD
CA610-701-124 RL610-701-124
Anti-IgG Donkey Polyclonal Antibody
Antibodies
Suitable for immunoblotting (western or dot blot), ELISA, electron microscopy and immunochemistry as well as other antibody-based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR