Anti-IgM µ Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
617-101-007 617-101-007
RL617-101-007EA 271.49 USD
CA617-101-007 RL617-101-007
Anti-IgM µ Goat Polyclonal Antibody
Antibodies
Anti-Monkey IgM (mu chain) is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR