Anti-CENPU Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-930 600-401-930
RL600-401-930EA 518.16 USD
RL600-401-930 CA600-401-930
Anti-CENPU Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA, western blotting and immunochemistry.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 65 kDa in size.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:2000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: CENPU
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR