Anti-CENPU Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-923 600-401-923
RL600-401-923EA 518.16 USD
RL600-401-923 CA600-401-923
Anti-CENPU Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 65 kDa in size corresponding to MLF1IP Protein by western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500- 1:2000;

Type: Primary
Antigen: CENPU
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR